5,5-Dimethyl-1,3,2-dioxathiane 2-oxide 95%

Supplier: Ambeed
Danger

A855906-25G A855906-5G
76654-432EA 56 USD
76654-432 76654-434
5,5-Dimethyl-1,3,2-dioxathiane 2-oxide 95%
5,5-Dimethyl-1,3,2-dioxathiane 2-oxide

Precaución: for Research Use Only

Formula: C₅H₁₀O₃S
MDL Number: MFCD25954041
CAS Number: 1003-85-6
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR