(E)-4-Chlorobut-2-enoic acid 95%

Supplier: Ambeed
Danger

A855251-100MG A855251-1G A855251-250MG A855251-5G
76654-424EA 143.23 USD
76654-424 76654-426 76654-428 76654-430
(E)-4-Chlorobut-2-enoic acid 95%
(E)-4-Chlorobut-2-enoic acid

Precaución: for Research Use Only

Formula: C₄H₅ClO₂
MW: 120.5343 g/mol
MDL Number: MFCD19228094
CAS Number: 26340-58-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR