2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A855089-25G A855089-5G
76654-416EA 58.35 USD
76654-416 76654-418
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid 97%
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
MDL Number: MFCD00006023
CAS Number: 59299-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR