1-Bromocyclopropane-1-carbonitrile 95%

Supplier: Ambeed
Warning

A854569-1G A854569-5G
76654-412EA 107.16 USD
76654-412 77573-710
1-Bromocyclopropane-1-carbonitrile 95%
1-Bromocyclopropane-1-carbonitrile

Precaución: for Research Use Only

Formula: C₄H₄BrN
MW: 145.99 g/mol
MDL Number: MFCD19688552
CAS Number: 1350746-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR