GIBH-130 98%

Supplier: Ambeed
Warning

Synonyms: (4-methyl-6-phenylpyridazin-3-yl)-(4-pyrimidin-2-ylpiperazin-1-yl)methanone, (4-Methyl-6-phenylpyridazin-3-yl)(4-(pyrimidin-2-yl)piperazin-1-yl)methanone

A854093-10MG A854093-1MG A854093-50MG A854093-5MG
76654-400EA 45.62 USD
76654-400 76654-402 76654-404 76654-406
GIBH-130 98%
GIBH-130

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₆O
MW: 360.42 g/mol
MDL Number: MFCD31544336
CAS Number: 1252608-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR