(1R,4R)-rel-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate 98%

Supplier: Ambeed
Warning

A854088-100MG A854088-1G A854088-250MG
76654-394EA 42.44 USD
76654-394 76654-396 76654-398
(1R,4R)-rel-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate 98%
(1R,4R)-rel-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
CAS Number: 1217975-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR