2,4,6-Trichloropyrido[3,4-d]pyrimidine 95%

Supplier: Ambeed
Warning

A853976-100MG A853976-250MG
76654-390EA 39 USD
76654-390 76654-392
2,4,6-Trichloropyrido[3,4-d]pyrimidine 95%
2,4,6-Trichloropyrido[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₂Cl₃N₃
MDL Number: MFCD31699745
CAS Number: 2143878-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR