2,5-Dimethylbenzene-1,4-dicarbaldehyde 95%

Supplier: Ambeed
Warning

A853347-250MG A853347-1G
76654-384EA 52 USD
76654-384 77784-234
2,5-Dimethylbenzene-1,4-dicarbaldehyde 95%
2,5-Dimethylbenzene-1,4-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00719462
CAS Number: 7044-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR