6-Methoxybenzo[d]oxazole 95%

Supplier: Ambeed
Warning

A852833-1G A852833-250MG A852833-100MG
76654-376EA 294.94 USD
76654-376 76654-378 77749-659
6-Methoxybenzo[d]oxazole 95%
6-Methoxybenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.1467 g/mol
MDL Number: MFCD26395367
CAS Number: 114980-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR