6-Chloro-2-methylpyridine-3-sulfonyl chloride 95%
Supplier: Ambeed
|
Danger
|
A852563-1G
A852563-25G
76654-374EA
43
USD
76654-374
77784-232
6-Chloro-2-methylpyridine-3-sulfonyl chloride 95%
6-Chloro-2-methylpyridine-3-sulfonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₅Cl₂NO₂S MW: 226.08 g/mol |
MDL Number:
MFCD08436477 CAS Number: 889944-74-5 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...