4-Hydroxybenzene-1-sulfonyl fluoride 95%

Supplier: Ambeed
Danger

A852371-100MG A852371-250MG A852371-50MG
76654-368EA 50 USD
76654-368 77784-230 77784-231
4-Hydroxybenzene-1-sulfonyl fluoride 95%
4-Hydroxybenzene-1-sulfonyl fluoride

Precaución: for Research Use Only

Formula: C₆H₅FO₃S
MDL Number: MFCD23110943
CAS Number: 368-87-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR