4-(Ethoxycarbonyl)oxane-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A851862-100MG A851862-250MG A851862-1G
76654-360EA 36 USD
76654-360 76654-362 77599-178
4-(Ethoxycarbonyl)oxane-4-carboxylic acid 95%
4-(Ethoxycarbonyl)oxane-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₅
MW: 202.2045 g/mol
MDL Number: MFCD24485511
CAS Number: 108630-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR