2-Methyl-4H-1-benzopyran-4-one 95%

Supplier: Ambeed
Warning

A851479-100MG A851479-1G A851479-250MG A851479-5G
76654-348EA 17 USD
76654-348 76654-350 76654-352 76654-354
2-Methyl-4H-1-benzopyran-4-one 95%
2-Methyl-4H-1-benzopyran-4-one

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MDL Number: MFCD00086982
CAS Number: 5751-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR