4-(6-(4-Methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)benzene-1,2-diamine trihydrochloride ≥97%

Supplier: Ambeed
Warning

A850530-100MG A850530-250MG A850530-50MG
76654-330EA 300.24 USD
76654-330 76654-332 76654-334
4-(6-(4-Methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)benzene-1,2-diamine trihydrochloride ≥97%
4-(6-(4-Methylpiperazin-1-yl)-1H-benzo[d]imidazol-2-yl)benzene-1,2-diamine trihydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₅Cl₃N₆
MW: 431.7903 g/mol
MDL Number: MFCD25977132
CAS Number: 1431697-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR