1-Phenylprop-2-yn-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A850288-1G A850288-250MG
76654-326EA 378 USD
76654-326 76654-328
1-Phenylprop-2-yn-1-amine hydrochloride 95%
1-Phenylprop-2-yn-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀ClN
MDL Number: MFCD00600875
CAS Number: 157022-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR