Sodium 2-(isoquinolin-1-yl)acetate 95%

Supplier: Ambeed
Warning

A850210-100MG A850210-1G A850210-250MG
76654-320EA 160 USD
76654-320 76654-322 76654-324
Sodium 2-(isoquinolin-1-yl)acetate 95%
Sodium 2-(isoquinolin-1-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₈NNaO₂
MW: 209.1765 g/mol
MDL Number: MFCD28118836
CAS Number: 1798728-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR