4-Fluoro-2-formylbenzoic acid 95%

Supplier: Ambeed
Warning

A850007-100MG A850007-1G A850007-250MG A850007-5G
76654-316EA 34 USD
76654-316 76654-318 77618-374 77749-657
4-Fluoro-2-formylbenzoic acid 95%
4-Fluoro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅FO₃
MDL Number: MFCD20658188
CAS Number: 1186047-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR