4-Bromo-N-methylpicolinamide 98%

Supplier: Ambeed
Warning

A849952-100MG A849952-1G A849952-250MG
76654-310EA 20 USD
76654-310 76654-312 76654-314
4-Bromo-N-methylpicolinamide 98%
4-Bromo-N-methylpicolinamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MDL Number: MFCD14702606
CAS Number: 1209459-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR