N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine xhydrochloride 99%

Supplier: Ambeed
Warning

A848717-100G A848717-10G A848717-25G A848717-500G A848717-1000G
76654-300EA 34 USD
76654-300 76654-302 76654-304 76654-306 77573-694
N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine xhydrochloride 99%
N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine xhydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈ClN₃
MW: 191.7016 g/mol
MDL Number: MFCD00012503
CAS Number: 7084-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR