4-(2,6-Difluorophenyl)piperidin-4-ol hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A848400-100MG
A848400-1G
A848400-250MG
76654-288EA
70.02
USD
76654-288
76654-290
76654-292
4-(2,6-Difluorophenyl)piperidin-4-ol hydrochloride 95%
4-(2,6-Difluorophenyl)piperidin-4-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₄ClF₂NO MW: 249.6848 g/mol |
MDL Number:
MFCD27500987 CAS Number: 1803595-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...