Sodium 6-fluoropyridine-2-sulfinate 95%

Supplier: Ambeed
Warning

A848340-100MG A848340-1G A848340-250MG
76654-282EA 142.17 USD
76654-282 76654-284 76654-286
Sodium 6-fluoropyridine-2-sulfinate 95%
Sodium 6-fluoropyridine-2-sulfinate

Precaución: for Research Use Only

Formula: C₅H₃FNNaO₂S
MW: 183.1360 g/mol
MDL Number: MFCD30536973
CAS Number: 2622208-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR