4-Methyl-1-phenyl-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A847792-100MG A847792-250MG
76654-274EA 163 USD
76654-274 76654-276
4-Methyl-1-phenyl-1H-pyrazole-3-carboxylic acid 95%
4-Methyl-1-phenyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD24480178
CAS Number: 241798-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR