4-Butoxy-2-chlorobenzoic acid 95%

Supplier: Ambeed
Warning

A847696-1G A847696-250MG
76654-270EA 152 USD
76654-270 76654-272
4-Butoxy-2-chlorobenzoic acid 95%
4-Butoxy-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MDL Number: MFCD00599178
CAS Number: 95736-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR