6-Amino-2-(butylamino)-9-((6-(2-(dimethylamino)ethoxy)pyridin-3-yl)methyl)-7,9-dihydro-8H-purin-8-one 98%

Supplier: Ambeed
Warning

Synonyms: DSR-6434

A846098-10MG A846098-1MG A846098-25MG A846098-50MG
76654-248EA 69 USD
76654-248 76654-250 76654-252 77623-042
6-Amino-2-(butylamino)-9-((6-(2-(dimethylamino)ethoxy)pyridin-3-yl)methyl)-7,9-dihydro-8H-purin-8-one 98%
6-Amino-2-(butylamino)-9-((6-(2-(dimethylamino)ethoxy)pyridin-3-yl)methyl)-7,9-dihydro-8H-purin-8-one

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₈O₂
MW: 400.48 g/mol
MDL Number: MFCD30182245
CAS Number: 1059070-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR