5-Ethynylpyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A845582-1G A845582-250MG A845582-5G
76654-244EA 108 USD
76654-244 76654-246 77552-584
5-Ethynylpyridine-3-carboxylic acid 95%
5-Ethynylpyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MDL Number: MFCD18207405
CAS Number: 1211533-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR