Methyl 2-bromo-3,3-dimethoxypropanoate 98%

Supplier: Ambeed
Danger

A844993-100MG A844993-1G A844993-250MG A844993-5G
76654-226EA 48.81 USD
76654-226 76654-228 76654-230 77599-170
Methyl 2-bromo-3,3-dimethoxypropanoate 98%
Methyl 2-bromo-3,3-dimethoxypropanoate

Precaución: for Research Use Only

Formula: C₆H₁₁BrO₄
MW: 227.0531 g/mol
MDL Number: MFCD22392188
CAS Number: 191330-98-0
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR