2,4-Dichlorobenzofuro[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A844556-1G A844556-5G
76654-224EA 175.05 USD
76654-224 77599-166
2,4-Dichlorobenzofuro[3,2-d]pyrimidine 97%
2,4-Dichlorobenzofuro[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₄Cl₂N₂O
MW: 239.06 g/mol
MDL Number: MFCD13190348
CAS Number: 160199-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR