5-Chloro-2-(2-(2-((3-(furan-3-yl)phenyl)amino)-2-oxoethoxy)acetamido)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: TM5441

A843529-100MG A843529-10MG A843529-1MG A843529-25MG A843529-2MG A843529-50MG A843529-5MG
76654-202EA 228.1 USD
76654-202 76654-204 76654-206 76654-208 76654-210 76654-212 76654-214
5-Chloro-2-(2-(2-((3-(furan-3-yl)phenyl)amino)-2-oxoethoxy)acetamido)benzoic acid 98%
5-Chloro-2-(2-(2-((3-(furan-3-yl)phenyl)amino)-2-oxoethoxy)acetamido)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₇ClN₂O₆
MW: 428.82 g/mol
MDL Number: MFCD31382187
CAS Number: 1190221-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR