2-(6-Chloropyridin-2-yl)propan-2-ol 97%

Supplier: Ambeed
Warning

A843407-100MG A843407-250MG
76654-198EA 30.77 USD
76654-198 76654-200
2-(6-Chloropyridin-2-yl)propan-2-ol 97%
2-(6-Chloropyridin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD19690108
CAS Number: 40472-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR