3,3,3-Trifluoropropane-1,2-diol 95%

Supplier: Ambeed
Danger

A843084-100MG A843084-1G A843084-250MG A843084-25G A843084-5G A843084-10G
76654-188EA 11 USD
76654-188 76654-190 76654-192 76654-194 76654-196 77623-024
3,3,3-Trifluoropropane-1,2-diol 95%
3,3,3-Trifluoropropane-1,2-diol

Precaución: for Research Use Only

Formula: C₃H₅F₃O₂
MDL Number: MFCD02183521
CAS Number: 431-39-0
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR