1,3,5-Tribromo-2,4,6-trifluorobenzene 97%

Supplier: Ambeed
Warning

A842242-100MG A842242-1G A842242-250MG A842242-5G
76654-178EA 8 USD
76654-178 76654-180 76654-182 76654-184
1,3,5-Tribromo-2,4,6-trifluorobenzene 97%
1,3,5-Tribromo-2,4,6-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆Br₃F₃
MDL Number: MFCD00192722
CAS Number: 2368-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR