11-(tert-Butoxy)-11-oxoundecanoic acid 97%
Supplier: Ambeed
|
Warning
|
A842145-100MG
A842145-10G
A842145-1G
A842145-250MG
A842145-25G
A842145-5G
76654-166EA
7
USD
76654-166
76654-168
76654-170
76654-172
76654-174
76654-176
11-(tert-Butoxy)-11-oxoundecanoic acid 97%
11-(tert-Butoxy)-11-oxoundecanoic acid
Precaución: for Research Use Only
Formula:
C₁₅H₂₈O₄ |
MDL Number:
MFCD30737824 CAS Number: 1789702-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...