(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-bromo-1H-indol-3-yl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Trp(5-Br)-OH

A841454-1G A841454-5G A841454-250MG
76654-154EA 120 USD
76654-154 76654-156 77784-196
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-bromo-1H-indol-3-yl)propanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-bromo-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₁BrN₂O₄
MW: 505.3600 g/mol
MDL Number: MFCD28166519
CAS Number: 460751-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR