Ethyl 1-amino-1H-pyrrole-3-carboxylate 95%

Supplier: Ambeed
Warning

A840770-100MG A840770-50MG A840770-1G A840770-250MG
76654-150EA 137.92 USD
76654-150 76654-152 77595-340 77618-354
Ethyl 1-amino-1H-pyrrole-3-carboxylate 95%
Ethyl 1-amino-1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
CAS Number: 941693-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR