(S)-2-Phenoxypropan-1-ol 95%

Supplier: Ambeed
Warning

A840470-100MG A840470-1G A840470-250MG
76654-144EA 84.88 USD
76654-144 76654-146 76654-148
(S)-2-Phenoxypropan-1-ol 95%
(S)-2-Phenoxypropan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.1900 g/mol
MDL Number: MFCD30164055
CAS Number: 87860-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR