Sm 324405 98%

Supplier: Ambeed
Warning

A839990-10MG A839990-1MG A839990-25MG A839990-50MG A839990-5MG
76654-134EA 59 USD
76654-134 76654-136 76654-138 76654-140 76654-142
Sm 324405 98%
Sm 324405

Precaución: for Research Use Only

Formula: C₁₉H₂₃N₅O₄
MDL Number: MFCD30182295
CAS Number: 677773-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR