(S)-Methyl 2-amino-3-cyclopropylpropanoate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A839574-1G
A839574-250MG
76654-124EA
108
USD
76654-124
76654-126
(S)-Methyl 2-amino-3-cyclopropylpropanoate hydrochloride 95%
(S)-Methyl 2-amino-3-cyclopropylpropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD28129824 CAS Number: 206438-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...