N-(Adamantan-1-yl)quinoxaline-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: NPS 2390

A839361-10MG A839361-1MG A839361-50MG A839361-5MG
76654-116EA 51.99 USD
76654-116 76654-118 76654-120 76654-122
N-(Adamantan-1-yl)quinoxaline-2-carboxamide 98%
N-(Adamantan-1-yl)quinoxaline-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₃O
MW: 307.3895 g/mol
MDL Number: MFCD05865239
CAS Number: 226878-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR