3,3'-Disulfanediylbis(propan-1-ol) 97%

Supplier: Ambeed
Danger

A838847-100MG A838847-10G A838847-1G A838847-250MG A838847-5G
76654-102EA 43.5 USD
76654-102 76654-104 76654-106 76654-108 77623-018
3,3'-Disulfanediylbis(propan-1-ol) 97%
3,3'-Disulfanediylbis(propan-1-ol)

Precaución: for Research Use Only

Formula: C₆H₁₄O₂S₂
MW: 182.3042 g/mol
MDL Number: MFCD01630941
CAS Number: 30453-21-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR