Ethyl 3-(3,4-difluorophenyl)-5-methyl-1H-pyrrole-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A838504-100MG
A838504-250MG
76654-098EA
88.06
USD
76654-098
76654-100
Ethyl 3-(3,4-difluorophenyl)-5-methyl-1H-pyrrole-2-carboxylate 97%
Ethyl 3-(3,4-difluorophenyl)-5-methyl-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₁₃F₂NO₂ MW: 265.2553 g/mol |
MDL Number:
MFCD31728058 CAS Number: 2288710-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...