1-Methyl-3-((methylamino)methyl)-1H-pyrazole-5-carbonitrile hydrochloride 98%

Supplier: Ambeed
Warning

A838499-1G A838499-250MG A838499-5G
76654-092EA 43.5 USD
76654-092 76654-094 76654-096
1-Methyl-3-((methylamino)methyl)-1H-pyrazole-5-carbonitrile hydrochloride 98%
1-Methyl-3-((methylamino)methyl)-1H-pyrazole-5-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₄
MW: 186.6420 g/mol
MDL Number: MFCD31697852
CAS Number: 1643141-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR