3-Benzyl-2-methoxyquinoline 98%

Supplier: Ambeed
Warning

A837533-1G A837533-100MG A837533-250MG
76654-086EA 154 USD
76654-086 77623-006 77784-186
3-Benzyl-2-methoxyquinoline 98%
3-Benzyl-2-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO
MDL Number: MFCD28156356
CAS Number: 1381767-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR