2-Bromo-5H-cyclopenta[b]pyridin-7(6H)-one 95%

Supplier: Ambeed
Warning

A837136-100MG A837136-1G A837136-250MG A837136-5G
76654-082EA 102 USD
76654-082 76654-084 77599-154 77749-636
2-Bromo-5H-cyclopenta[b]pyridin-7(6H)-one 95%
2-Bromo-5H-cyclopenta[b]pyridin-7(6H)-one

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD18255917
CAS Number: 1256823-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR