tert-Butyl 4-(4-amino-2-fluorophenyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A836838-1G A836838-250MG
76654-080EA 135 USD
76654-080 77749-635
tert-Butyl 4-(4-amino-2-fluorophenyl)piperidine-1-carboxylate 97%
tert-Butyl 4-(4-amino-2-fluorophenyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₃FN₂O₂
MW: 294.3644 g/mol
MDL Number: MFCD15071912
CAS Number: 188975-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR