(R)-Methyl 2-amino-3-(2-fluoro-4-methylphenyl)propanoate hydrochloride 95%

Supplier: Ambeed
Warning

A836456-100MG A836456-250MG
76654-074EA 45 USD
76654-074 76654-076
(R)-Methyl 2-amino-3-(2-fluoro-4-methylphenyl)propanoate hydrochloride 95%
(R)-Methyl 2-amino-3-(2-fluoro-4-methylphenyl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClFNO₂
MDL Number: MFCD24434472
CAS Number: 2682097-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR