(R)-2-Amino-2-(4-iodophenyl)ethan-1-ol hydrochloride 95%

Supplier: Ambeed
Warning

A836432-100MG A836432-250MG
76654-070EA 114.58 USD
76654-070 76654-072
(R)-2-Amino-2-(4-iodophenyl)ethan-1-ol hydrochloride 95%
(R)-2-Amino-2-(4-iodophenyl)ethan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClINO
MW: 299.5365 g/mol
MDL Number: MFCD18375925
CAS Number: 2829292-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR