2,6-Dimethoxyanilino(oxo)acetic acid 98%

Supplier: Ambeed
Danger

A835648-250MG A835648-5G A835648-1G
76654-058EA 20.16 USD
76654-058 76654-060 77573-660
2,6-Dimethoxyanilino(oxo)acetic acid 98%
2,6-Dimethoxyanilino(oxo)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₅
MW: 225.1980 g/mol
MDL Number: MFCD29905054
CAS Number: 2097273-59-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR