(E)-6-Chloro-3-(3-(5-fluoropyridin-3-yl)acryloyl)-4-phenylquinolin-2(1H)-one 98%
Supplier: Ambeed
|
Warning
|
A835082-1MG
A835082-25MG
A835082-5MG
76654-052EA
13
USD
76654-052
76654-054
76654-056
(E)-6-Chloro-3-(3-(5-fluoropyridin-3-yl)acryloyl)-4-phenylquinolin-2(1H)-one 98%
(E)-6-Chloro-3-(3-(5-fluoropyridin-3-yl)acryloyl)-4-phenylquinolin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₂₃H₁₄ClFN₂O₂ |
CAS Number:
2205871-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...