2-((4-(Cyclopropanecarbonyl)piperazin-1-yl)methyl)benzo[d]isothiazol-3(2H)-one 98%

Supplier: Ambeed
Warning

A834854-10MG A834854-1MG A834854-25MG A834854-50MG
76654-038EA 76 USD
76654-038 76654-040 76654-042 76654-044
2-((4-(Cyclopropanecarbonyl)piperazin-1-yl)methyl)benzo[d]isothiazol-3(2H)-one 98%
2-((4-(Cyclopropanecarbonyl)piperazin-1-yl)methyl)benzo[d]isothiazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₂S
MDL Number: MFCD07849042
CAS Number: 877963-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR