(E)-2-(Thiophen-2-yl)ethyl 2-cyano-3-(3,4-dihydroxyphenyl)acrylate 99%

Supplier: Ambeed
Warning

A834096-10MG A834096-1MG A834096-50MG
76654-024EA 22 USD
76654-024 76654-026 76654-028
(E)-2-(Thiophen-2-yl)ethyl 2-cyano-3-(3,4-dihydroxyphenyl)acrylate 99%
(E)-2-(Thiophen-2-yl)ethyl 2-cyano-3-(3,4-dihydroxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₄S
MDL Number: MFCD00894731
CAS Number: 132465-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR